Learn More
Fine Science Tools Moria Mini Perforated Spoon, Tip: 8 mm, 10 cm
SDP
Catalog No. NC1197733
Encompass
Individual Item Name: Moria Mini Perforated Spoon - 10cm/ Tip Diameter: 8mm/ Spoon Depth: 1mm/ Alloy: Inox/ Length: 10cm
Catalog No. NC1197733 Supplier Fine Science Tools Supplier No. 1037019
Individual Item Name: Moria Mini Perforated Spoon - 10cm/ Tip Diameter: 8mm/ Spoon Depth: 1mm/ Alloy: Inox/ Length: 10cm