Learn More
Fine Science Tools MORIA SPOON,8MM DIA X 2MM DEEP
SDP
Catalog No. NC1820772
Encompass
Moria SpoonTip Diameter:8mmSpoon Depth:2mmAlloy / Material:InoxLength:12.5cm
Catalog No. NC1820772 Supplier Fine Science Tools Supplier No. 1032108
Moria SpoonTip Diameter:8mmSpoon Depth:2mmAlloy / Material:InoxLength:12.5cm