Learn More
Fine Science Tools MORIA VESSEL CLAMPS STRAIGHT
SDP
Catalog No. NC9040646
Encompass
Product: Moria Vessel Clamp - Straight/33mm/ Shape: Straight/ Jaw Dimensions: 13 x 1.6mm/ Clamping Pressure: Light/ Alloy: Inox/ Length: 33mm/ Serrated: Yes/ Unit: Each
Catalog No. NC9040646 Supplier Fine Science Tools Supplier No. 1832011
Product: Moria Vessel Clamp - Straight/33mm/ Shape: Straight/ Jaw Dimensions: 13 x 1.6mm/ Clamping Pressure: Light/ Alloy: Inox/ Length: 33mm/ Serrated: Yes/ Unit: Each