Learn More
Medchemexpress LLC Moricizine Hydrochloride | 29560-58-5 | 98.0% | 5 MG
SDP
Catalog No. 5000408753
Encompass_Preferred
Moricizine Hydrochloride (EN 313) is an orally active Class I antiarrhythmic agent. It works by decreasing the maximum rate of phase 0 depolarization, increasing the rates of phase 2 and 3 repolarization, and decreasing both the action potential duration and the effective refractory period.
  • Orally active class I antiarrhythmic agent
  • Decreases the maximum rate of phase 0 depolarization
  • Increases rates of phase 2 and 3 repolarization
  • Decreases action potential duration
  • Decreases effective refractory period
  • Target: Cytochrome P450
  • Pathway: Metabolic enzyme/protease
Catalog No. 50-004-08753 Supplier Medchemexpress LLC Supplier No. HYB0615A5MG
Moricizine Hydrochloride (EN 313) is an orally active Class I antiarrhythmic agent. It works by decreasing the maximum rate of phase 0 depolarization, increasing the rates of phase 2 and 3 repolarization, and decreasing both the action potential duration and the effective refractory period.
  • Orally active class I antiarrhythmic agent
  • Decreases the maximum rate of phase 0 depolarization
  • Increases rates of phase 2 and 3 repolarization
  • Decreases action potential duration
  • Decreases effective refractory period
  • Target: Cytochrome P450
  • Pathway: Metabolic enzyme/protease