Learn More
Medchemexpress LLC Morin | 480-16-0 | 97.4% | 302.24 g·mol⁻1 | C15H10O7 | 10 MG
SDP
Catalog No. 5000296500
Encompass_Preferred
Morin is a plant-derived flavonoid used in research to modulate protein tyrosine phosphatase activity and oxidative stress pathways. It is reported to inhibit PTP1B (IC50 ≈ 15 μM), activate the insulin receptor, and influence apoptotic signaling. Molecular weight 302.24 g·mol⁻1; typical purity ~97.4%.
  • PTP1B inhibitor with reported IC50 ≈ 15 μM.
  • Modulates reactive oxygen species and apoptosis pathways.
  • Useful for metabolic disease and cancer biology studies.
  • Provided in small research quantities suitable for in vitro assays.
  • Powder form is stable under recommended frozen storage conditions.
Catalog No. 50-002-96500 Supplier Medchemexpress LLC Supplier No. HYN062110MG
Morin is a plant-derived flavonoid used in research to modulate protein tyrosine phosphatase activity and oxidative stress pathways. It is reported to inhibit PTP1B (IC50 ≈ 15 μM), activate the insulin receptor, and influence apoptotic signaling. Molecular weight 302.24 g·mol⁻1; typical purity ~97.4%.
  • PTP1B inhibitor with reported IC50 ≈ 15 μM.
  • Modulates reactive oxygen species and apoptosis pathways.
  • Useful for metabolic disease and cancer biology studies.
  • Provided in small research quantities suitable for in vitro assays.
  • Powder form is stable under recommended frozen storage conditions.