Learn More
Sigma Aldrich Fine Chemicals Biosciences Morin hydrate for microscopy (Fl.), for the determination of Al, Be, Zn, Ga, In, Sc, 1-2 mol/mol water | 654055-01-3 | MFCD00017307 | 10G
Catalog No. 501867886
Encompass
Morin hydrate for microscopy (Fl.), for the determination of Al, Be, Zn, Ga, In, Sc, 1-2 mol/mol water | Purity: >=85% (HPLC) | 654055-01-3 | MFCD00017307 | 10G
Catalog No. 50-186-7886 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 6987010G
Morin hydrate for microscopy (Fl.), for the determination of Al, Be, Zn, Ga, In, Sc, 1-2 mol/mol water | Purity: >=85% (HPLC) | 654055-01-3 | MFCD00017307 | 10G