Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Morin, MP Biomedicals™

Catalog No. icn15573305 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15573305 5 g
ICN15573310 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15573305 Supplier MP Biomedicals, Inc Supplier No. 0215573305
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 480-16-0
Molecular Formula C15H10O7
Molecular Weight (g/mol) 302.238
InChI Key YXOLAZRVSSWPPT-UHFFFAOYSA-N
Synonym morin, aurantica, calico yellow, al-morin, toxylon pomiferum, bois d,arc, morin hydrate, 2',3,4',5,7-pentahydroxyflavone, osage orange, osage orange extract
PubChem CID 5281670
ChEBI CHEBI:75092
IUPAC Name 2-(2,4-dihydroxyphenyl)-3,5,7-trihydroxychromen-4-one
SMILES C1=CC(=C(C=C1O)O)C2=C(C(=O)C3=C(C=C(C=C3O2)O)O)O

Specifications

Color Yellow to Brown
Quantity 5 g
Formula Weight 302.2
Physical Form Powder
Chemical Name or Material Morin
EINECSNumber 207-542-9
Recommended Storage Store at 2° to 8°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.