Learn More
Medchemexpress LLC HY-15781A 5mg Medchemexpress, Morinidazole (R enantiomer) CAS:898230-59-6 Purity:>98%
SDP
Catalog No. 501969956
Encompass_Preferred
Medchemexpress, HY-15781A 5mg Morinidazole (R enantiomer) CAS:898230-59-6 Morinidazole R enantiomer is the R-enantiomer of Morinidazole. Morinidazole is a new 5-nitroimidazole class antimicrobial agent. Morinidazole R enantiomer is the less active enantiomer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9956 Supplier Medchemexpress LLC Supplier No. HY15781A5MG
Medchemexpress, HY-15781A 5mg Morinidazole (R enantiomer) CAS:898230-59-6 Morinidazole R enantiomer is the R-enantiomer of Morinidazole. Morinidazole is a new 5-nitroimidazole class antimicrobial agent. Morinidazole R enantiomer is the less active enantiomer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.