Learn More
MSC Diversey Neutral Disinfectant Cleaner, Quaternary-Based, Non-Alkaline, 4 EA
Catalog No. 502866217
Encompass
This quaternary-based non-alkaline disinfectant offers excellent cleaning properties, suitable for healthcare, hotels, schools, and restaurants. It is a bactericide and virucide with a 10-minute contact time, effectively killing MRSA and VRE, and meeting bloodborne pathogen standards.
  • Disinfectant cleaner that is bactericidal, virucidal, and kills MRSA and VRE
  • Hospital use disinfectant with a 10-minute contact time that meets bloodborne pathogen standards for decontaminating blood and bodily fluids
  • General cleaner and disinfectant for larger areas such as operating rooms and patient care facilities
Catalog No. 50-286-6217 Supplier MSC Supplier No. 03037991
This quaternary-based non-alkaline disinfectant offers excellent cleaning properties, suitable for healthcare, hotels, schools, and restaurants. It is a bactericide and virucide with a 10-minute contact time, effectively killing MRSA and VRE, and meeting bloodborne pathogen standards.
  • Disinfectant cleaner that is bactericidal, virucidal, and kills MRSA and VRE
  • Hospital use disinfectant with a 10-minute contact time that meets bloodborne pathogen standards for decontaminating blood and bodily fluids
  • General cleaner and disinfectant for larger areas such as operating rooms and patient care facilities