Learn More
Cayman Chemical Mornteltartrate 100mg
SDP
Catalog No. 502995599
Encompass_Preferred
A positive allosteric modulator of nAChRs; enhances channel gating of the α3β2 nAChR subtype
Catalog No. 50-299-5599 Supplier Cayman Chemical Supplier No. 22902100 MG
A positive allosteric modulator of nAChRs; enhances channel gating of the α3β2 nAChR subtype