Learn More
Apexbio Technology LLC Moroxydine HCl 3160-91-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000092922
Encompass_Preferred
Moroxydine hydrochloride (3160-91-6) is a small-molecule antiviral agent designed to interfere with viral nucleic acid replication thereby inhibiting the synthesis of viral DNA and RNA Moroxydine hydrochloride exerts its biological activity primarily through inhibition of viral nucleic acid replication mechanisms In in vitro studies Moroxydine hydrochloride demonstrates inhibitory activity against influenza virus herpes simplex virus (HSV) measles virus varicella-zoster virus (VZV) mumps virus hepatitis C virus (HCV) and Grass Carp Reovirus (GCRV) Reported antiviral effects typically involve IC50 values in the micromolar range though precise IC50 values depend on the viral strain and experimental conditions Based on these pharmacological properties Moroxydine hydrochloride holds research potential in antiviral mechanism studies and drug screening assays
Catalog No. 50-000-92922 Supplier Apexbio Technology LLC Supplier No. B19805.1
Moroxydine hydrochloride (3160-91-6) is a small-molecule antiviral agent designed to interfere with viral nucleic acid replication thereby inhibiting the synthesis of viral DNA and RNA Moroxydine hydrochloride exerts its biological activity primarily through inhibition of viral nucleic acid replication mechanisms In in vitro studies Moroxydine hydrochloride demonstrates inhibitory activity against influenza virus herpes simplex virus (HSV) measles virus varicella-zoster virus (VZV) mumps virus hepatitis C virus (HCV) and Grass Carp Reovirus (GCRV) Reported antiviral effects typically involve IC50 values in the micromolar range though precise IC50 values depend on the viral strain and experimental conditions Based on these pharmacological properties Moroxydine hydrochloride holds research potential in antiviral mechanism studies and drug screening assays