Learn More
Medchemexpress LLC Moroxydine (hydrochloride) | 3160-91-6 | 99.4% | 10 G
SDP
Catalog No. 5000409245
Encompass_Preferred
Moroxydine hydrochloride (ABOB hydrochloride) is a broad-spectrum agent with multi-antiviral activities against both DNA and RNA viruses. These include influenza virus, herpes simplex, varicella zoster, measles, mumps disease, and hepatitis C virus. It demonstrates excellent antiviral activity with low cytotoxicity to cells infected by dsRNA viruses like grass carp reovirus (GCRV) and large DNA viruses such as giant salamander iridovirus (GSIV).
  • Blocks GCRV-induced cytopathic effects
  • Removes nucleocapsids in ctenopharyngodon idella kidney (CIK) cells
  • Maintains normal morphological structure of CIK cells
  • Significantly inhibits apoptosis
  • Inhibits caspase 3 activity and Bax expression
  • Down-regulates Bcl-2 levels
Catalog No. 50-004-09245 Supplier Medchemexpress LLC Supplier No. HYB0420A10G
Moroxydine hydrochloride (ABOB hydrochloride) is a broad-spectrum agent with multi-antiviral activities against both DNA and RNA viruses. These include influenza virus, herpes simplex, varicella zoster, measles, mumps disease, and hepatitis C virus. It demonstrates excellent antiviral activity with low cytotoxicity to cells infected by dsRNA viruses like grass carp reovirus (GCRV) and large DNA viruses such as giant salamander iridovirus (GSIV).
  • Blocks GCRV-induced cytopathic effects
  • Removes nucleocapsids in ctenopharyngodon idella kidney (CIK) cells
  • Maintains normal morphological structure of CIK cells
  • Significantly inhibits apoptosis
  • Inhibits caspase 3 activity and Bax expression
  • Down-regulates Bcl-2 levels