Learn More
Molecular Dimensions MORPHEUS BUFFER SYSTEM 1 100ML
SDP
Catalog No. NC9059906
Encompass_Preferred
Protein Crystallography. 100mL: Contains untitrated 1M Imidazole and untitrated 1M MES. Mix together according to Buffer Composition Table (contained with kit) to achieve a range of pH's to optimize with. All our stock reagents are made with ultra-purified water and filter sterilized.
Catalog No. NC9059906 Supplier Molecular Dimensions Supplier No. MD2100100
Protein Crystallography. 100mL: Contains untitrated 1M Imidazole and untitrated 1M MES. Mix together according to Buffer Composition Table (contained with kit) to achieve a range of pH's to optimize with. All our stock reagents are made with ultra-purified water and filter sterilized.