Learn More
Molecular Dimensions MORPHEUS BUFFER SYSTEM 2, 100M
SDP
Catalog No. NC0720600
Encompass
Protein Crystallography. 100mL: Contains untitrated 1M Sodium HEPES and untitrated 1M MOPS. Mix together according to Buffer Composition Table (contained with kit) to achieve a range of pH's to optimize with. All our stock reagents are made with ultra-purified water and filter sterilized.
Catalog No. NC0720600 Supplier Molecular Dimensions Supplier No. MD2100101
Protein Crystallography. 100mL: Contains untitrated 1M Sodium HEPES and untitrated 1M MOPS. Mix together according to Buffer Composition Table (contained with kit) to achieve a range of pH's to optimize with. All our stock reagents are made with ultra-purified water and filter sterilized.