Learn More
Molecular Dimensions Morpheus - Buffer System 3, 250mL
SDP
Catalog No. NC0529654
Encompass_Preferred
Protein Crystallography. 250mL: Contains untitrated 1M Tris and untitrated 1M BICINE. Mix together according to Buffer Composition Table (contained with kit) to achieve a range of pH's to optimize with.All our stock reagents are made with ultra-purified water and filter sterilized.
Catalog No. NC0529654 Supplier Molecular Dimensions Supplier No. MD2250102
Protein Crystallography. 250mL: Contains untitrated 1M Tris and untitrated 1M BICINE. Mix together according to Buffer Composition Table (contained with kit) to achieve a range of pH's to optimize with.All our stock reagents are made with ultra-purified water and filter sterilized.