Learn More
Molecular Dimensions MORPHEUS EDO_P8K MIX
SDP
Catalog No. NC0488377
Encompass
Protein Crystallography. 100mL: Precipitant Mix 2 (EDO_P8K Mix) from Morpheus Contains a mixture of Ethylene glycol and PEG 8000 made with ultra-purified water.
Catalog No. NC0488377 Supplier Molecular Dimensions Supplier No. MD210082
Protein Crystallography. 100mL: Precipitant Mix 2 (EDO_P8K Mix) from Morpheus Contains a mixture of Ethylene glycol and PEG 8000 made with ultra-purified water.