Learn More
Molecular Dimensions MORPHEUS ETHYLENE GLYCOLS MIX,
SDP
Catalog No. NC0720604
Encompass
Protein Crystallography. 100mL: Ethylene glycols Mix from Morpheus screen. Contains a mixture of diethylene glycol, triethylene glycol, tetraethylene glycol, and pentaethylene glycol.
Catalog No. NC0720604 Supplier Molecular Dimensions Supplier No. MD210074
Protein Crystallography. 100mL: Ethylene glycols Mix from Morpheus screen. Contains a mixture of diethylene glycol, triethylene glycol, tetraethylene glycol, and pentaethylene glycol.