Learn More
Molecular Dimensions MORPHEUS II HT-96
SDP
Supplier:  Molecular Dimensions MD192
Encompass_Preferred
Protein Crystallography. MORPHEUS II screen HT-96 (96 x 1.1mL):Targeted incorporation of 35 low- molecular weight PDB ligands into 96 conditions.
Catalog No. NC0919295