Learn More
Molecular Dimensions MORPHEUS MONOSACCHARIDES MIX
SDP
Catalog No. NC0488376
Encompass
Protein Crystallography. 100mL: Monosaccharides Mix from Morpheus screen. Contains D-glucose, D-mannose, D-galactose, L-fucose, D-xylose, N-acetyl-D-glucosamine. All our stock reagents are made with ultra-purified water and filter sterilized.
Catalog No. NC0488376 Supplier Molecular Dimensions Supplier No. MD210075
Protein Crystallography. 100mL: Monosaccharides Mix from Morpheus screen. Contains D-glucose, D-mannose, D-galactose, L-fucose, D-xylose, N-acetyl-D-glucosamine. All our stock reagents are made with ultra-purified water and filter sterilized.