Learn More
Molecular Dimensions MORPHEUS P550MME-P20K (100 ML)
SDP
Catalog No. NC0109106
Encompass
Protein Crystallography. 100mL: Precipitant Mix 1 (PEG500MME_P20K Mix) from Morpheus Contains a mixture of PEG 500 MME and PEG 20,000 made with ultra-purified water.
Catalog No. NC0109106 Supplier Molecular Dimensions Supplier No. MD210081
Protein Crystallography. 100mL: Precipitant Mix 1 (PEG500MME_P20K Mix) from Morpheus Contains a mixture of PEG 500 MME and PEG 20,000 made with ultra-purified water.