Learn More
Cayman Chemical Morphine ELISA Standard, 1 ea
SDP
Catalog No. 502942251
Encompass_Preferred
Morphine ELISA Standard has been tested and formulated to work exclusively with Cayman’s Morphine ELISA Kit (Item No. 501940).
Catalog No. 50-294-2251 Supplier Cayman Chemical Supplier No. 4019441 EA
Morphine ELISA Standard has been tested and formulated to work exclusively with Cayman’s Morphine ELISA Kit (Item No. 501940).