Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Morpholine, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019649 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9649 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9649 Supplier MilliporeSigma Supelco Supplier No. 9017910MLF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Morpholines are systemic compounds that exert both protective and curative actions against fungal diseases.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 110-91-8
Ignition Point 590°F
Vapor Pressure 7 mmHg (20°C)
Vapor Density 3 (vs. air)
Density 0.996 g/mL (at 25°C)
Flash Point 31°C
Physical Form Neat
Linear Formula C4H9 NO
MDL Number MFCD00005972
Melting Point -7°C to -5°C
Boiling Point 129°C
Synonym Tetrahydro-1,4-oxazine
Type Analytical Standard
Quantity 10 mL
Formula Weight 87.12
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.