Learn More
Medchemexpress LLC Morpholine-d4 hydrochloride | 1115228-35-7 | 127.61 | 1 G
SDP
Catalog No. 5000454973
Encompass_Preferred
Morpholine-d4 hydrochloride is the deuterium labeled Morpholine hydrochloride. It is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-54973 Supplier Medchemexpress LLC Supplier No. HYW012821SA1G
Morpholine-d4 hydrochloride is the deuterium labeled Morpholine hydrochloride. It is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.