Learn More
Medchemexpress LLC Morpholine-d4 hydrochloride | 1115228-35-7 | 127.61 | 100 MG
SDP
Catalog No. 5000454936
Encompass_Preferred
Morpholine-d4 hydrochloride is the deuterium labeled Morpholine hydrochloride. This compound can be used as a tracer or as an internal standard for quantitative analysis. It is suitable for analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-54936 Supplier Medchemexpress LLC Supplier No. HYW012821SA100MG
Morpholine-d4 hydrochloride is the deuterium labeled Morpholine hydrochloride. This compound can be used as a tracer or as an internal standard for quantitative analysis. It is suitable for analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS