Learn More
Medchemexpress LLC Morpholine-d4 hydrochloride | 1115228-35-7 | 127.61 | 250 MG
SDP
Catalog No. 5000451311
Encompass_Preferred
Morpholine-d4 hydrochloride is the deuterium labeled Morpholine hydrochloride. It is for research use only and not sold to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-51311 Supplier Medchemexpress LLC Supplier No. 250MGHYW012821SA
Morpholine-d4 hydrochloride is the deuterium labeled Morpholine hydrochloride. It is for research use only and not sold to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS