Learn More
Medchemexpress LLC Morpholine-d4 hydrochloride | 1115228-35-7 | 98.0% | 127.61 | 500 MG
SDP
Catalog No. 5000454820
Encompass_Preferred
Morpholine-d4 hydrochloride is the deuterium labeled Morpholine hydrochloride. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-54820 Supplier Medchemexpress LLC Supplier No. HYW012821SA500MG
Morpholine-d4 hydrochloride is the deuterium labeled Morpholine hydrochloride. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS