Learn More
Crescent Chemical Co Inc MORPHOLINOETHANE SULFONIC ACID
SDP
Catalog No. 50247886
Encompass_Preferred
Morpholinoethane sulfonic acid analytical grade pKa 20 = 6.15. Buffering substance for biochemistry and molecular biology (1). Physical constants (2). Size - 500G Storage Conditions - +15 °C TO +30 °C Catalog Number - 29834.04 CAS 4432-31-9
Catalog No. 50-247-886 Supplier Crescent Chemical Co Inc Supplier No. SE29834.04
Morpholinoethane sulfonic acid analytical grade pKa 20 = 6.15. Buffering substance for biochemistry and molecular biology (1). Physical constants (2). Size - 500G Storage Conditions - +15 °C TO +30 °C Catalog Number - 29834.04 CAS 4432-31-9