Learn More
Crescent Chemical Co Inc MORPHOLINOPROPANE SULFONICACID
SDP
Supplier:  Crescent Chemical Co Inc SE29836.03
Encompass_Preferred
Morpholinopropane sulfonic acid analytical grade pKa 20 = 7.2. Buffering substance for biochemistry, molecular and cell biology. Buffer component for isoelectric focussing and SDS PAGE. Substitute for HCO3-/CO2-buffer in cell culture for muscle studies. Size - 500G Sto
Catalog No. 50-247-888