Learn More
Crescent Chemical Co Inc MORPHOLINOPROPANE SULFONICACID
SDP
Catalog No. 50247887
Encompass_Preferred
Morpholinopropane sulfonic acid analytical grade pKa 20 = 7.2. Buffering substance for biochemistry, molecular and cell biology. Buffer component for isoelectric focussing and SDS PAGE. Substitute for HCO3-/CO2-buffer in cell culture for muscle studies. Size - 100G Sto
Catalog No. 50-247-887 Supplier Crescent Chemical Co Inc Supplier No. SE29836.02
Morpholinopropane sulfonic acid analytical grade pKa 20 = 7.2. Buffering substance for biochemistry, molecular and cell biology. Buffer component for isoelectric focussing and SDS PAGE. Substitute for HCO3-/CO2-buffer in cell culture for muscle studies. Size - 100G Sto