Learn More
Electron Microscopy Sciences EMS Morphology Stage Micrometer, Model IAM-1
SDP
Catalog No. 5034575
Encompass_Preferred
Ideally for morphological calibration where two dimensional shapes have to be measured precisely.  Standard includes a set of eight test plates of various shapes, positions, angles and sizes. It also includes a linear measurement scale for calibrating your imaging system. Overall size: 1" x 3" (25 mm x 75 mm). Positive image / Chrome on glass (PG)
Catalog No. 50-345-75 Supplier Electron Microscopy Sciences Supplier No. 602161PG
Ideally for morphological calibration where two dimensional shapes have to be measured precisely.  Standard includes a set of eight test plates of various shapes, positions, angles and sizes. It also includes a linear measurement scale for calibrating your imaging system. Overall size: 1" x 3" (25 mm x 75 mm). Positive image / Chrome on glass (PG)