Learn More
Molecular Dimensions MORPHUS MPDP1KP3350 MIX 100ML
SDP
Catalog No. NC0608559
Encompass
Protein Crystallography. 100mL: Precipitant Mix 4 (MPD_P1K_P3350 Mix) from Morpheus Contains a mixture of MPD, PEG 1000 and PEG 3350 made with ultra-purified water.
Catalog No. NC0608559 Supplier Molecular Dimensions Supplier No. MD210084
Protein Crystallography. 100mL: Precipitant Mix 4 (MPD_P1K_P3350 Mix) from Morpheus Contains a mixture of MPD, PEG 1000 and PEG 3350 made with ultra-purified water.