Learn More
Moore Medical MorTan™ Morgan™ Lens Delivery Set
Description
Requires
MorTan™ Lens (Sold separately)
Specialized IV delivery set which can be attached to two Morgan lenses.
- Saves time and money by eliminating the need to use two separate IV setups
- Rate of flow may be controlled and irrigation to just one eye is possible with the included clamps
Specifications
Specifications
| Product Type | Lens Delivery Set |
| For Use With (Equipment) | Specialized for I.V |
| For Use With (Application) | Controlled flow rate |