Learn More
Cole-Parmer Cole-Parmer Mortar and Pestle Set, porcelain, 50 mL
Supplier:  Cole-Parmer 6310061
Encompass_Preferred
Mortar features lip and spout, and is glazed on the outside (except the bottom). Oversized pestle is also glazed (except the grinding surface). Mortar and pestle are sold as a set. Autoclavable
Catalog No. NC1696242