Learn More
Fritsch Milling And Sizing Inc Mortar P-0 Hardened Steel
SDP
Catalog No. 501534352
Encompass
General material specification:
Material: hardened steel - 1.2080
Abrasion resistance: good
Suitable for: medium-hard, brittle samples, suitable for cryogenic grinding
Catalog No. 50-153-4352 Supplier Fritsch Milling And Sizing Inc Supplier No. 40.0120.09
General material specification:
Material: hardened steel - 1.2080
Abrasion resistance: good
Suitable for: medium-hard, brittle samples, suitable for cryogenic grinding