Learn More
Fritsch Milling And Sizing Inc Mortar P-0 Hardmetal Tungsten Carbide
SDP
Catalog No. 501534351
Encompass
General material specification:
Material: hardmetal tungsten carbide - WC - GC10
Abrasion resistance: very good
Suitable for: hard, abrasive samples, very suitable for cryogenic grinding
Catalog No. 50-153-4351 Supplier Fritsch Milling And Sizing Inc Supplier No. 40.0110.08
Add to Cart
Add to Cart
Request a Quote
General material specification:
Material: hardmetal tungsten carbide - WC - GC10
Abrasion resistance: very good
Suitable for: hard, abrasive samples, very suitable for cryogenic grinding