Learn More
Gilson Company Inc Mortar & Pestle
Catalog No. NC0894890
Encompass_Preferred
Mortar & Pestle is made of heavy porcelain. Mortar is 5in (127mm), Dia. and has 320ml capacity. The mortar is glazed except for the sample contact area and comes with a small porcelain pestle.
Catalog No. NC0894890 Supplier Gilson Company Inc Supplier No. HM109
Mortar & Pestle is made of heavy porcelain. Mortar is 5in (127mm), Dia. and has 320ml capacity. The mortar is glazed except for the sample contact area and comes with a small porcelain pestle.