Learn More
Cole-Parmer Cole-Parmer Mortar and Pestle Set, Porcelain; 300 mL, Each
Catalog No. NC0414482 Shop All Cole-Parmer Products
Encompass_Preferred
Mortars are glazed on the outside and pestles are glazed to grinding surface. Features pouring spout for easy pouring. Both mortar and pestle are sold as a set.
Catalog No. NC0414482 Supplier Cole-Parmer Supplier No. 6310058
Mortars are glazed on the outside and pestles are glazed to grinding surface. Features pouring spout for easy pouring. Both mortar and pestle are sold as a set.