Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Mortar for Tapered Tissue Grinder, 5 ml, 13 mm x 93 mm Mortor, 220 mm OAL
SDP

Catalog No. 501900354
Encompass_Preferred

Replacement parts for Tapered tissue grinder.
Size: 5 ml
Mortor: 13 mm x 93 mm
OAL: 220 mm

Catalog No. 50-190-0354 Supplier Electron Microscopy Sciences Supplier No. 6479672
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Replacement parts for Tapered tissue grinder.
Size: 5 ml
Mortor: 13 mm x 93 mm
OAL: 220 mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.