Learn More
Alkali Scientific Mortexer™  Combo-head Vortexer 115V
SDP
Catalog No. 502336151
Encompass_Preferred
The Mortexer™ is MORE than your standard vortex mixer. The unique design of the Multi-Head (included) features a traditional cup head for general purpose mixing, plus holds up to eight microcentrifuge tubes. The drive system is balanced with Q-Drive technology, providing a smooth vortexing action, while minimizing noise and excessive vibration. This results in quieter, more efficient and longer lasting operation. Increased speed, up to 3400 rpm produces an instant vortex of even the largest sample sizes, including full 50mL tubes. Other features include: "no-walk" suction feet, cold room/incubator compatibility and spill-proof electronic controls. Two year warranty. Unique Multi-Head for general purpose vortexing PLUS holds up to eight microtubes (Patent Pending)Powerful motor for INSTANT vortexingContinuous or "touch" operationQ-Drive dynamic balancing systemVariable speed from 200 to 3400 rpmWide variety of accessories
Catalog No. 50-233-6151 Supplier Alkali Scientific Supplier No. RS7002
The Mortexer™ is MORE than your standard vortex mixer. The unique design of the Multi-Head (included) features a traditional cup head for general purpose mixing, plus holds up to eight microcentrifuge tubes. The drive system is balanced with Q-Drive technology, providing a smooth vortexing action, while minimizing noise and excessive vibration. This results in quieter, more efficient and longer lasting operation. Increased speed, up to 3400 rpm produces an instant vortex of even the largest sample sizes, including full 50mL tubes. Other features include: "no-walk" suction feet, cold room/incubator compatibility and spill-proof electronic controls. Two year warranty. Unique Multi-Head for general purpose vortexing PLUS holds up to eight microtubes (Patent Pending)Powerful motor for INSTANT vortexingContinuous or "touch" operationQ-Drive dynamic balancing systemVariable speed from 200 to 3400 rpmWide variety of accessories