Learn More
Medchemexpress LLC HY-N0622 5mg Medchemexpress, Morusin CAS:62596-29-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN06225MG
Encompass_Preferred
Medchemexpress, HY-N0622 5mg Morusin CAS:62596-29-6 Morusin is a prenylated flavonoid isolated from M. australis with various biological activities, such as antitumor, antioxidant, and anti-bacteria property. Morusin could inhibit NF-κB and STAT3 activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3645