Learn More
Aviva Systems Biology Corp MOS Antibody - middle region (ARP56631_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56631_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MOS. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MOS. Alternate names: MGC119962, MGC119963, MSVApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61932