Learn More
Bioworld MOSAIC HEPATITIS C VIRUS NS
Supplier:  Bioworld 220605651
Encompass_Preferred
MOSAIC HEPATITIS C VIRUS NS4 GENOTYPE-2(RECOMBINANT): Recombinant Mosaic Hepatitis C Virus nonstructural protein NS4 Genotype 2 Source:E.coli Description : Artificial mosaic polypeptide composite constructed from diagnostically relev...
Catalog No. 50-253-665