Learn More
Apexbio Technology LLC Mosapride Citrate 112885-42-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000095553
Encompass_Preferred
Mosapride Citrate (CAS 112885-42-4) is a small molecule that functions as a selective agonist of the 5-hydroxytryptamine 4 (5-HT ) receptor and an antagonist of the 5-HT receptor with negligible activity at dopaminergic D adrenergic and as well as 5-HT and 5-HT receptors By modulating serotonin receptor activity it enhances gastrointestinal motility and accelerates gastric emptying Mosapride Citrate is widely used in research investigating serotonergic signaling pathways gut motility regulation and potential therapeutic approaches for gastrointestinal disorders
Catalog No. 50-000-95553 Supplier Apexbio Technology LLC Supplier No. A13345.1
Mosapride Citrate (CAS 112885-42-4) is a small molecule that functions as a selective agonist of the 5-hydroxytryptamine 4 (5-HT ) receptor and an antagonist of the 5-HT receptor with negligible activity at dopaminergic D adrenergic and as well as 5-HT and 5-HT receptors By modulating serotonin receptor activity it enhances gastrointestinal motility and accelerates gastric emptying Mosapride Citrate is widely used in research investigating serotonergic signaling pathways gut motility regulation and potential therapeutic approaches for gastrointestinal disorders