Learn More
Medchemexpress LLC Mosapride citrate | 112885-42-4 | 99.9% | 1 ML
SDP
Catalog No. 5000405762
Encompass_Preferred
Mosapride citrate is an orally active gastroenterokinetic compound. It acts as a 5HT4 agonist and a CYP inducer. It exhibits a concentration-dependent inhibitory effect on Kv4.3, with an IC50 value of 15.2 μM. Mosapride citrate is used in the study of gastrointestinal diseases.
  • Orally active gastroenterokinetic compound
  • 5HT4 agonist
  • CYP inducer
  • Inhibitory effect on Kv4.3
  • Useful for studying gastrointestinal diseases
Catalog No. 50-004-05762 Supplier Medchemexpress LLC Supplier No. HYB0189A10MM/1ML
Mosapride citrate is an orally active gastroenterokinetic compound. It acts as a 5HT4 agonist and a CYP inducer. It exhibits a concentration-dependent inhibitory effect on Kv4.3, with an IC50 value of 15.2 μM. Mosapride citrate is used in the study of gastrointestinal diseases.
  • Orally active gastroenterokinetic compound
  • 5HT4 agonist
  • CYP inducer
  • Inhibitory effect on Kv4.3
  • Useful for studying gastrointestinal diseases