Learn More
Medchemexpress LLC Mosapride citrate dihydrate | 636582-62-2 | 99.6% | 25 MG
SDP
Catalog No. 5000451006
Encompass_Preferred
Mosapride citrate dihydrate is a gastroprokinetic agent that acts as a 5-hydroxytryptamine4 receptor agonist. It has been extensively used in research related to various gastrointestinal disorders. This compound also potently inhibits Kv4.3 in a concentration-dependent manner, with an IC50 value of 15.2 μM. In vivo, it specifically stimulates upper gastrointestinal motility.
  • Acts as a 5-hydroxytryptamine4 receptor agonist
  • Used in research for gastrointestinal disorders
  • Potently inhibits Kv4.3
  • Selectively stimulates upper gastrointestinal motility in vivo
Catalog No. 50-004-51006 Supplier Medchemexpress LLC Supplier No. 25MGHYB0189B
Mosapride citrate dihydrate is a gastroprokinetic agent that acts as a 5-hydroxytryptamine4 receptor agonist. It has been extensively used in research related to various gastrointestinal disorders. This compound also potently inhibits Kv4.3 in a concentration-dependent manner, with an IC50 value of 15.2 μM. In vivo, it specifically stimulates upper gastrointestinal motility.
  • Acts as a 5-hydroxytryptamine4 receptor agonist
  • Used in research for gastrointestinal disorders
  • Potently inhibits Kv4.3
  • Selectively stimulates upper gastrointestinal motility in vivo