Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Moschamine | 193224-22-5 | 98.1% | 352.38 g/mol | C20H20N2O4 | 250 MG
SDP

Catalog No. 5000229007
Encompass_Preferred

Moschamine is an endogenous metabolite supplied for research use that inhibits excessive mitochondrial superoxide production. It is provided as a solid for in vitro research applications and is not for human or clinical use.

  • Inhibits excessive mitochondrial superoxide production.
  • High reported purity of 98.1%.
  • Molecular weight 352.38 g/mol and formula C20H20N2O4.
  • Soluble in DMSO at approximately 100 mg/mL; ultrasonic assistance may be required.
  • Stable as a solid at room temperature (3 years); in solvent: -80°C (2 years), -20°C (1 year).
  • Available in multiple pack sizes, including a 250 mg vial for convenient handling.

Catalog No. 50-002-29007 Supplier Medchemexpress LLC Supplier No. HY118824250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Moschamine is an endogenous metabolite supplied for research use that inhibits excessive mitochondrial superoxide production. It is provided as a solid for in vitro research applications and is not for human or clinical use.

  • Inhibits excessive mitochondrial superoxide production.
  • High reported purity of 98.1%.
  • Molecular weight 352.38 g/mol and formula C20H20N2O4.
  • Soluble in DMSO at approximately 100 mg/mL; ultrasonic assistance may be required.
  • Stable as a solid at room temperature (3 years); in solvent: -80°C (2 years), -20°C (1 year).
  • Available in multiple pack sizes, including a 250 mg vial for convenient handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.