Learn More
TARGETMOL CHEMICALS INC MOSLOFLAVONE 5MG
SDP
Catalog No. 502579733
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. MOSLOFLAVONE showed promising anti-inflammatory activity via inhibition of TNF-(alpha) and IL-1(beta) with IC50 values of 0.71 uM and 7.8 uM respectively. purity: 98%
Catalog No. 50-257-9733 Supplier TARGETMOL CHEMICALS INC Supplier No. T57795MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. MOSLOFLAVONE showed promising anti-inflammatory activity via inhibition of TNF-(alpha) and IL-1(beta) with IC50 values of 0.71 uM and 7.8 uM respectively. purity: 98%