Learn More
MSC Mosmatic Rotary Cleaner, 30 in, 4000 PSI, 1 ea
Catalog No. 502820014
Encompass
The Mosmatic FL-CR 750 Commercial Surface Cleaner is designed for efficient, high-pressure cleaning of large surfaces. Constructed from durable stainless steel, this unit provides powerful and consistent performance in commercial and industrial settings, with minimal maintenance requirements.
  • Extra-wide cleaning path with 30 in diameter for fast and efficient cleaning
  • Adjustable low-pressure system for accurate detergent use with chemical application
  • Designed for long-term use with minimal maintenance
  • High-performance stainless steel swivel with reinforced ball bearings and dual spring carbide seals
  • Precision rotor arms support up to 4000 PSI for uniform and streak-free cleaning
  • Four 2 1/4 in stainless steel casters for smooth, stable movement
Catalog No. 50-282-0014 Supplier MSC Supplier No. 17297292
The Mosmatic FL-CR 750 Commercial Surface Cleaner is designed for efficient, high-pressure cleaning of large surfaces. Constructed from durable stainless steel, this unit provides powerful and consistent performance in commercial and industrial settings, with minimal maintenance requirements.
  • Extra-wide cleaning path with 30 in diameter for fast and efficient cleaning
  • Adjustable low-pressure system for accurate detergent use with chemical application
  • Designed for long-term use with minimal maintenance
  • High-performance stainless steel swivel with reinforced ball bearings and dual spring carbide seals
  • Precision rotor arms support up to 4000 PSI for uniform and streak-free cleaning
  • Four 2 1/4 in stainless steel casters for smooth, stable movement