Learn More
MSC Moso Natural Air Purifier, 300 gram, Odor Control, 1 ea
Catalog No. 502818306
Encompass
This air purifying bag effectively eliminates odors and harmful pollutants, while also regulating humidity in damp environments. Operating without batteries or wires, it continuously absorbs impurities from the air.
  • Absorbs odors and harmful pollutants
  • Controls humidity in damp, musty spaces
  • Chemical-free, fragrance-free, and non-toxic
  • Lasts up to two years with routine rejuvenation
  • Freestanding design allows for flexible placement
  • Disposed charcoal promotes plant growth
Catalog No. 50-281-8306 Supplier MSC Supplier No. 14203350
This air purifying bag effectively eliminates odors and harmful pollutants, while also regulating humidity in damp environments. Operating without batteries or wires, it continuously absorbs impurities from the air.
  • Absorbs odors and harmful pollutants
  • Controls humidity in damp, musty spaces
  • Chemical-free, fragrance-free, and non-toxic
  • Lasts up to two years with routine rejuvenation
  • Freestanding design allows for flexible placement
  • Disposed charcoal promotes plant growth