Learn More
Aviva Systems Biology Corp MOSPD3 Antibody - C-terminal region (ARP44910_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44910_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MOSPD3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human MOSPD3. Alternate names: CDS3, NET30ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58934